• USA Home
  • 70011 - Multielement atomic spectroscopy standard solution VII

EMAIL THIS PAGE TO A FRIEND
70011 Sigma-Aldrich

Multielement atomic spectroscopy standard solution VII

in hydrochloric acid

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Materials Science,
shelf life   limited shelf life, expiry date on the label
composition   Au, 10 mg/L
  Ge, 10 mg/L
  Pt, 10 mg/L
  Sn, 10 mg/L
  Ti, 10 mg/L
  Zr, 10 mg/L
concentration   in hydrochloric acid
application(s)   ICP/AAS: suitable
format   multi-component solution

Description

Preparation Note

prepared with high purity salts and hydrochloric acid (~1 M)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?