• USA Home
  • 70021 - Myeloperoxidase from human leukocytes

EMAIL THIS PAGE TO A FRIEND
70021 Sigma-Aldrich

Myeloperoxidase from human leukocytes

lyophilized, powder, white, ≥50 U/mg protein

Synonym: MPO, Peroxidase, myelo

Purchase

Description

General description

lyophilized from 20 mM sodium acetate buffer, pH 6,0

Other Notes

Review

Same product — same quality — now Sigma-Aldrich brand

Unit Definition

1 U corresponds to the amount of enzyme which will produce an increase in absorption (A470) of 1.0 per minute at pH 7.0 and 25°C, calculated from the initial rate of reaction using guaiacol (Cat. No. 50880) as substrate; total reaction volume: 3.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

M6908 Myeloperoxidase from human leukocytes, lyophilized powder, ≥50 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?