• USA Home
  • 70025 - Myoglobin from equine skeletal muscle

EMAIL THIS PAGE TO A FRIEND
70025 Sigma-Aldrich

Myoglobin from equine skeletal muscle

≥90% (GE)

Synonym: Myoglobin from horse skeletal muscle

Purchase

Properties

assay   ≥90% (GE)
mol wt   Mr ~17600
  mol wt ~17 kDa(lit.)
impurities   ≤10% water
solubility   H2O: 10 mg/mL clear, red to red-brown
cation traces   Fe: 0.25-0.35%
absorbance ratio   A280/525 nm ~4.5
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Sales restrictions may apply

Biochem/physiol Actions

Myoglobin is critical to skeletal muscle O2 supply at near-maximum oxygen demand, and prevents anoxia by maintaining PO2 above levels needed to support mitochondrial function.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M0630 Myoglobin from equine skeletal muscle, 95-100%, essentially salt-free, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?