• USA Home
  • 70054 - AQUANAL-professional ALKA-M-Photometer

EMAIL THIS PAGE TO A FRIEND
70054 Sigma-Aldrich

AQUANAL-professional ALKA-M-Photometer

for the photometric determination of m-value as calcium carbonate range 5-200 mg/L

Purchase

Description

Packaging

100 tablets

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
RTECS 
AU8400000
Flash Point(F) 
384.8 °F
Flash Point(C) 
196 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?