• USA Home
  • 70056 - AQUANAL-professional ALKA-P-Photometer
EMAIL THIS PAGE TO A FRIEND
70056 Sigma-Aldrich
AQUANAL-professional ALKA-P-Photometer
for the photometric determination of p-value as calcium carbonate range: 5-300 mg/L
Purchase
Description
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
100 tablets
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?