• USA Home
  • 70057 - AQUANAL-professional Ammonia No. 1
EMAIL THIS PAGE TO A FRIEND
70057 Sigma-Aldrich
AQUANAL-professional Ammonia No. 1
determination of ammonium; use with : ammonium no. 2
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
measuring range   0.02-1 mg/L, (photometric, Spectro 1000)
  0.02-2.0 mg/L, (photometric, Spectro 3)
Description
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
100 tablets
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?