• USA Home
  • 70070 - AQUANAL-professional Acidifying GP
EMAIL THIS PAGE TO A FRIEND
70070 Sigma-Aldrich
AQUANAL-professional Acidifying GP
determination of chlorine, to be used with 70068
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
measuring range   5-200 mg/L
Description
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
100 tablets
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?