• USA Home
  • 70076 - AQUANAL-professional Iron No. 1 LR
EMAIL THIS PAGE TO A FRIEND
70076 Sigma-Aldrich
AQUANAL-professional Iron No. 1 LR
determination of iron; use with : iron no. 2 LR
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
measuring range   0.01-1 mg/L
Description
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
100 tablets
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?