• USA Home
  • 70086 - AQUANAL-professional Copper No. 2
EMAIL THIS PAGE TO A FRIEND
70086 Sigma-Aldrich
AQUANAL-professional Copper No. 2
determination of copper; use with : copper no. 1
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
measuring range   0.05-5 mg/L
Description
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
100 tablets
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?