• USA Home
  • 70107 - AQUANAL-professional Nitrate tube test

EMAIL THIS PAGE TO A FRIEND
70107 Sigma-Aldrich

AQUANAL-professional Nitrate tube test

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
measuring range   0.5-60 mg/L

Description

Packaging

24 reaction tubes, 1 blank tube

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
WGK Germany 
3
Flash Point(F) 
86 °F
Flash Point(C) 
30 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?