• USA Home
  • 70113 - AQUANAL-professional Vario H Chlorine Free (DPD)
EMAIL THIS PAGE TO A FRIEND
70113 Sigma-Aldrich
AQUANAL-professional Vario H Chlorine Free (DPD)
determination of chlorine
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
measuring range   0-5 mg/L, (Cl2)
Description
Analysis Note
method: N,N-Diethyl-p-phenylenediamine (DPD)
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
pack contains 100 powder bags (each makes 25 mL)
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?