EMAIL THIS PAGE TO A FRIEND
70140 Sigma-Aldrich

Sodium myristate

≥98.0% (NT)

Synonym: Myristic acid sodium salt, Tetradecanoic acid sodium salt

  • CAS Number 822-12-8

  • Linear Formula CH3(CH2)12COONa

  • Molecular Weight 250.35

  •  Beilstein Registry Number 3575157

  •  EC Number 212-487-9

  •  MDL number MFCD00042653

  •  PubChem Substance ID 329762286

Purchase

Description

Application

Sodium myristate was used for the N-myristoylation of recoverin .

Biochem/physiol Actions

Myristic acid is commonly added via a covalent linkage to the N-terminal glycine of many eukaryotic and viral proteins, a process called myristoylation. Myristoylation enables proteins to bind to cell membranes and facilitates protein-protein interactions. Myristolyation of proteins affect many cellular functions and thus has implications in health and disease .

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M8005 Sodium myristate, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?