EMAIL THIS PAGE TO A FRIEND
70154 Sigma-Aldrich

(−)-cis-Myrtanol

analytical standard

Synonym: (1S,2R)-6,6-Dimethylbicyclo[3.1.1]heptane-2-methanol, (1S,2R)-10-Pinanol

  • CAS Number 51152-12-6

  • Empirical Formula (Hill Notation) C10H18O

  • Molecular Weight 154.25

  •  Beilstein Registry Number 2552096

  •  EC Number 257-018-9

  •  MDL number MFCD00066424

  •  PubChem Substance ID 329762299

Purchase

Properties

Related Categories Additional Standards, Alcohols, Aliphatic, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   LDWAIHWGMRVEFR-CIUDSAMLSA-N
assay   ≥98.5% (sum of enantiomers, GC)
form   neat
optical activity   [α]20/D −19.5±1°, neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.492
bp   127 °C/22 mmHg(lit.)
density   0.977 g/mL at 20 °C(lit.)
format   neat
storage temp.   2-8°C

Description

Other Notes

Preparation of a chiral reducing agent

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
154.4 °F
Flash Point(C) 
68 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?