EMAIL THIS PAGE TO A FRIEND
70155 Sigma-Aldrich

(−)-trans-Myrtanol

purum, ≥98.0% (sum of enantiomers, GC)

Synonym: (1S,2S)-10-Pinanol, (1S,2S)-6,6-Dimethylbicyclo[3.1.1]heptane-2-methanol

  • CAS Number 53369-17-8

  • Empirical Formula (Hill Notation) C10H18O

  • Molecular Weight 154.25

  •  Beilstein Registry Number 2322989

  •  EC Number 258-499-8

  •  MDL number MFCD00001348

  •  PubChem Substance ID 24886062

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   LDWAIHWGMRVEFR-VGMNWLOBSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D −30±2°, neat
refractive index   n20/D 1.490
bp   232-236 °C(lit.)
density   0.973 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NA 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
186.8 °F
Flash Point(C) 
86 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?