• USA Home
  • 70218 - AQUANAL-professional Vario H Nitrite-10
EMAIL THIS PAGE TO A FRIEND
70218 Sigma-Aldrich
AQUANAL-professional Vario H Nitrite-10
determination of nitrite
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
measuring range   0-0.2 mg/L, (NO2-N)
Description
Analysis Note
method: diazotization
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
100 powder packs for 10 mL sample volume
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?