• USA Home
  • 70224 - (+)-Ethyl (S)-2-hydroxy-4-oxo-4-phenylbutyrate

EMAIL THIS PAGE TO A FRIEND
70224 Sigma-Aldrich

(+)-Ethyl (S)-2-hydroxy-4-oxo-4-phenylbutyrate

purum, ≥97.0% (sum of enantiomers, HPLC)

  • CAS Number 243658-52-8

  • Empirical Formula (Hill Notation) C12H14O4

  • Molecular Weight 222.24

  •  Beilstein Registry Number 2695594

  •  MDL number MFCD03490502

  •  PubChem Substance ID 329762356

Purchase

Properties

InChI Key   HEWAMPODKFTBJZ-NSHDSACASA-N
grade   purum
assay   ≥97.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D +5.5±0.5°, c = 1% in chloroform
optical purity   enantiomeric ratio: ≥99:1 (HPLC)
mp   35-38 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?