• USA Home
  • 70244 - AQUANAL-professional Vario H Sulfate
EMAIL THIS PAGE TO A FRIEND
70244 Sigma-Aldrich
AQUANAL-professional Vario H Sulfate
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
measuring range   0-70 mg/L, (SO4)
Description
Analysis Note
method: barium chloride
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
100 powder packs for 25 mL sample volume
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
UN 3316 6.1/PG 3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?