Synonym: Potassium hexacyanoferrate(III), Red prussiate
CAS Number 13746-66-2
Linear Formula K3Fe(CN)6
EC Number 237-323-3
MDL number MFCD00011392
Popular Documents: Specification Sheet (PDF)
| assay | 99% |
| form | powder |
| particle size | <10 μm |
Live Chat and Frequently Asked Questions
are available for this Product.
50, 250 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?