• USA Home
  • 70270 - 2-Naphthalenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
70270 Sigma-Aldrich

2-Naphthalenesulfonyl chloride

puriss., ≥99.0% (AT)

  • CAS Number 93-11-8

  • Empirical Formula (Hill Notation) C10H7ClO2S

  • Molecular Weight 226.68

  •  Beilstein Registry Number 641898

  •  EC Number 202-219-9

  •  MDL number MFCD00004087

  •  PubChem Substance ID 329762395

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonyl Halides, Sulfur Compounds More...
InChI Key   OPECTNGATDYLSS-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (AT)
ign. residue   ≤0.05%
bp   147.7 °C/0.6 mmHg(lit.)
mp   74-76 °C(lit.)
  74-76 °C

Description

Other Notes

Amino protecting reagent which gives more labile derivatives than tosyl; 2-nasyl is selectively cleaved in the presence of tosyl by Mg/MeOH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

133361 2-Naphthalenesulfonyl chloride, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?