• USA Home
  • 70290 - Sodium 2-naphthalenesulfonate

EMAIL THIS PAGE TO A FRIEND
70290 Sigma-Aldrich

Sodium 2-naphthalenesulfonate

technical, ≥90%

Synonym: 2-Naphthalenesulfonic acid sodium salt

  • CAS Number 532-02-5

  • Empirical Formula (Hill Notation) C10H7NaO3S

  • Molecular Weight 230.22

  •  Beilstein Registry Number 3659024

  •  EC Number 208-523-8

  •  MDL number MFCD00064186

  •  PubChem Substance ID 24886006

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acids, Sulfur Compounds More...
InChI Key   YWPOLRBWRRKLMW-UHFFFAOYSA-M
grade   technical
assay   ≥90%

Description

Other Notes

Used for ion-pair chromatography of amines and amino acid derivatives with UV-detection

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
QK3678000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?