• USA Home
  • 70385 - 2,3-Naphtho-15-crown-5

EMAIL THIS PAGE TO A FRIEND
70385 Sigma-Aldrich

2,3-Naphtho-15-crown-5

Selectophore, ≥99.0%

  • CAS Number 17454-47-6

  • Empirical Formula (Hill Notation) C18H22O5

  • Molecular Weight 318.36

  •  Beilstein Registry Number 1651900

  •  MDL number MFCD00068674

  •  PubChem Substance ID 24886076

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, Crown Ethers, Synthetic Reagents
grade   Selectophore
InChI Key   BDTDDXDRCOLVNJ-UHFFFAOYSA-N
assay   ≥99.0% (HPLC)
  ≥99.0%
mp   116-118 °C

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Potassium-selective ionophore

Same product — same quality — now Sigma-Aldrich brand

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

99373 Potassium ionophore I - cocktail B, Selectophore
99311 Potassium ionophore I - cocktail A, Selectophore
60396 Potassium ionophore IV, Selectophore, function tested
60397 Potassium ionophore III, Selectophore, function tested
60401 Potassium ionophore II, Selectophore, function tested
60403 Potassium ionophore I, Selectophore, function tested

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?