EMAIL THIS PAGE TO A FRIEND
70440 Sigma-Aldrich

1-Naphthol

puriss., for fluorescence, ≥99.0% (GC)

Synonym: α-Naphthol, 1-Hydroxynaphthalene

  • CAS Number 90-15-3

  • Linear Formula C10H7OH

  • Molecular Weight 144.17

  •  Beilstein Registry Number 1817321

  •  EC Number 201-969-4

  •  MDL number MFCD00003930

Purchase

Properties

Related Categories Biochemicals and Reagents, Detection, Fluorescent Enzyme Substrates, Fluorescent Enzyme Substrates for Spectroscopy, Fluorescent Probes, Labels, Particles and Stains More...
grade   for fluorescence
vapor density   4.5 (120 °C, vs air)
vapor pressure   1 mmHg ( 94 °C)
InChI Key   KJCVRFUGPWSIIH-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
autoignition temp.   1007 °F
expl. lim.   5 %
ign. residue   ≤0.05%
bp   278-280 °C(lit.)
mp   94-96 °C(lit.)
  95-97 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

1-Naphthol (Naphthalen-1-ol) may be used as a standard for the NMR, FT-IR and mass spectrometric characterization of impurities present in the Duloxetine Hydrochloride active pharmaceutical ingredient (API).3

General description

1-Naphthol is the hydroxyl derivative of naphthalene. It forms hydrogen bonded complexes with H2O, CH3OH, NH3 and ND3 in gas phase. Hydrogen-bond dissociation energy of these complexes has been evaluated. Its sorption by biochars (obtained from orange peels, having pyrolytic temperatures in the range of 150-700°C and designated as OP150-OP700) has been studied. It has been identified as an impurity in the active pharmaceutical ingredient (API) of duloxetine hydrochloride.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN2811 - class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
1
RTECS 
QL2800000
Flash Point(F) 
257 °F
Flash Point(C) 
125 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

N1000 1-Naphthol, ReagentPlus®, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?