EMAIL THIS PAGE TO A FRIEND
70452 Sigma-Aldrich

2-Naphthol

purum, ≥98.0% (GC)

Synonym: β-Naphthol, 2-Hydroxynaphthalene

Purchase

Properties

Related Categories Building Blocks, C9 to C20+, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
vapor density   4.97 (vs air)
vapor pressure   10 mmHg ( 145.5 °C)
InChI Key   JWAZRIHNYRIHIV-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
impurities   ≤1% 1-naphthol
bp   285-286 °C(lit.)
mp   120-122 °C(lit.)
  120-122 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
QL2975000
Flash Point(F) 
307.4 °F
Flash Point(C) 
153 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

130109 2-Naphthol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?