• USA Home
  • 70471 - Naphthol AS phosphate

EMAIL THIS PAGE TO A FRIEND
70471 Sigma-Aldrich

Naphthol AS phosphate

for histology

Synonym: 3-Hydroxy-2-naphthanilide phosphate

  • CAS Number 13989-98-5

  • Empirical Formula (Hill Notation) C17H14NO5P

  • Molecular Weight 343.27

  •  Beilstein Registry Number 2772810

  •  EC Number 237-789-8

  •  MDL number MFCD00036182

  •  PubChem Substance ID 57652250

Purchase

Properties

Related Categories Acid Phosphatase, Alkaline Phosphatase, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates,
grade   for histology
InChI Key   KVIYXIWBXOQZDN-UHFFFAOYSA-N
storage temp.   2-8°C

Description

Other Notes

Substrate for the histochemical location of phosphatases

Substrates

Histochemical substrate for acid and alkaline phosphatase.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?