• USA Home
  • 70642 - Flask adapter for HPLC
EMAIL THIS PAGE TO A FRIEND
70642 Sigma-Aldrich
Flask adapter for HPLC
PTFE/FEP
Purchase
Description
General description
HPLC Adapter suitable for all bottles with DIN 32 thread. Adaptation for 2 HPLC-tubes with 1/16 iin. outer diameter
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?