• USA Home
  • 70644 - Silica gel on TLC Al foils

EMAIL THIS PAGE TO A FRIEND
70644 Sigma-Aldrich

Silica gel on TLC Al foils

silica gel matrix, L × W 5 cm × 10 cm

Purchase

Properties

Related Categories Aluminum TLC Plates: Silica matrix, Aluminum TLC Plates: without fluorescent indicator, Analytical/Chromatography, LPLC/TLC/Paper Chromatography, TLC Plates, Aluminum foil backing,
material   aluminum support
  silica gel matrix
feature   binder Polymeric
  fluorescent indicator: no
mfr. no.   Sigma-Aldrich
application(s)   thin layer chromatography (TLC): suitable
L × W   5 cm × 10 cm
layer thickness   200 μm
particle size   8.0-12.0 μm
pore size   60 Å medium pore diameter

Description

General description

• Outstanding wettability (even for 100% aqueous visualisation reagents)
• Easy and precise cutting (no flaking of silica)
• Excellent separation efficiency and reproducibility from batch to batch

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is widely used as layer material for TLC. Plaster of Paris is generally used as binding agent to hold silica gel firmly to the TLC plates. Silica gel is applied as sorbent to the aluminium foils.

Application

Silica gel on TLC Alu foils may be used to perform thin layer chromatography.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?