• USA Home
  • 70693 - (S)-(−)-1-(1-Naphthyl)ethanol

EMAIL THIS PAGE TO A FRIEND
70693 Sigma-Aldrich

(S)-(−)-1-(1-Naphthyl)ethanol

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym: (S)-(−)-α-Methyl-1-naphthalenemethanol

  • CAS Number 15914-84-8

  • Empirical Formula (Hill Notation) C12H12O

  • Molecular Weight 172.22

  •  Beilstein Registry Number 2045009

  •  MDL number MFCD00077831

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   CDRQOYRPWJULJN-VIFPVBQESA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D −78±2°, c = 1% in methanol
optical purity   enantiomeric ratio: ≥99:1 (HPLC)
mp   45-47.5 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?