• USA Home
  • 70711 - (R)-(+)-1-(1-Naphthyl)ethylamine

EMAIL THIS PAGE TO A FRIEND
70711 Sigma-Aldrich

(R)-(+)-1-(1-Naphthyl)ethylamine

purum, ≥98.0% (sum of enantiomers, GC)

Synonym: (R)-(+)-α-Methyl-1-naphthalenemethylamine

Purchase

Properties

Related Categories Amines, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   RTCUCQWIICFPOD-SECBINFHSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D +53±3°, c = 2% in ethanol
optical purity   enantiomeric ratio: ≥97.5:2.5 (GC)
color   brown
refractive index   n20/D 1.623(lit.)
  n20/D 1.623
bp   153 °C/11 mmHg(lit.)
density   1.067 g/mL at 20 °C(lit.)

Description

Other Notes

Reagent for the resolution of acids by salt formation; Starting material for (R)-1-(1-naphthyl)ethyl isocyanate

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?