• USA Home
  • 70721 - N-(1-Naphthyl)ethylenediamine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
70721 Sigma-Aldrich

N-(1-Naphthyl)ethylenediamine dihydrochloride

purum, ACS reagent, ≥97.0% (calc. based on dry substance, AT)

Synonym: 2-(1-Naphthylamino)ethylamine dihydrochloride

  • CAS Number 1465-25-4

  • Linear Formula C10H7NHCH2CH2NH2 · 2HCl

  • Molecular Weight 259.17

  •  Beilstein Registry Number 3707471

  •  EC Number 215-981-2

  •  MDL number MFCD00012556

Purchase

Properties

grade   ACS reagent
InChI Key   MZNYWPRCVDMOJG-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (calc. based on dry substance, AT)
ign. residue   ≤0.1%
mp   194-198 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
KV5330000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?