• USA Home
  • 71176 - Sodium ionophore I - cocktail A

EMAIL THIS PAGE TO A FRIEND
71176 Sigma-Aldrich

Sodium ionophore I - cocktail A

Selectophore

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophore Cocktails More...
grade   Selectophore
composition   2-nitrophenyl octyl ether (o-NPOE), 89.5 wt. %
  sodium ionophore (ETH 227), 10.0 wt. %
  sodium tetraphenylborate, 0.5 wt. %

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Ready-to-use ion-selective liquid for the preparation of microelectrodes in micropipettes.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?