• USA Home
  • 71177 - Sodium standard for AAS

EMAIL THIS PAGE TO A FRIEND
71177 Sigma-Aldrich

Sodium standard for AAS

ready-to-use, traceable to BAM, in H2O

Synonym: Sodium chloride solution

  • CAS Number 7647-14-5

  • Linear Formula NaCl

  • Molecular Weight 58.44

  •  Beilstein Registry Number 3534976

  •  MDL number MFCD00003477

  •  PubChem Substance ID 329763087

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Matrix Selection,
InChI Key   FAPWRFPIFSIZLT-UHFFFAOYSA-M
shelf life   limited shelf life, expiry date on the label
concentration   in H2O
  1.000 g/L Na
application(s)   ICP/AAS: suitable
format   single component solution

Description

Preparation Note

prepared with NaCl and H2O

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?