EMAIL THIS PAGE TO A FRIEND
71260 Sigma-Aldrich

Sodium amide

purum, ≥95.0% (T), crystalline

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Inorganic Bases, Synthetic Reagents More...
InChI Key   ODZPKZBBUMBTMG-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (T)
form   crystalline
bp   400 °C(lit.)
mp   210 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with water liberates toxic gas., May form explosive peroxides.
RIDADR 
UN 1390 4.3 / PGII
WGK Germany 
2
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?