EMAIL THIS PAGE TO A FRIEND
71445 Sigma-Aldrich

Sodium decyl sulfate

for surfactant analysis, ≥99.0% (T)

Synonym: Decyl sulfate sodium salt

  • CAS Number 142-87-0

  • Linear Formula CH3(CH2)9OSO3Na

  • Molecular Weight 260.33

  •  Beilstein Registry Number 3576860

  •  EC Number 205-568-5

  •  MDL number MFCD00041881

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Reagents for Surfactant Analysis
grade   for surfactant analysis
InChI Key   XZTJQQLJJCXOLP-UHFFFAOYSA-M
assay   ≥99.0% (T)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
WS7625000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?