• USA Home
  • 71481 - Sodium diethyldithiocarbamate trihydrate

EMAIL THIS PAGE TO A FRIEND
71481 Sigma-Aldrich

Sodium diethyldithiocarbamate trihydrate

≥97.0% (NT)

Synonym: Cupral, Diethyldithiocarbamic acid sodium salt

  • CAS Number 20624-25-3

  • Linear Formula (C2H5)2NCSSNa · 3H2O

  • Molecular Weight 225.31

  •  Beilstein Registry Number 3920507

  •  EC Number 205-710-6

  •  MDL number MFCD00150617

  •  PubChem Substance ID 329763316

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocarbonyl Compounds More...
vapor density   5.9 (vs air)
InChI Key   WWGXHTXOZKVJDN-UHFFFAOYSA-M
assay   ≥97.0% (NT)
mp   95-98.5 °C(lit.)

Description

Biochem/physiol Actions

Inhibits induction of macrophage nitric oxide synthase.

Application

Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO2 has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
EZ6550000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D3506 Sodium diethyldithiocarbamate trihydrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?