• USA Home
  • 71506 - Sodium phosphate monobasic monohydrate

EMAIL THIS PAGE TO A FRIEND
71506 Sigma-Aldrich

Sodium phosphate monobasic monohydrate

purum p.a., ≥99.0% (T)

Synonym: Monosodium phosphate, Sodium dihydrogen phosphate monohydrate

Purchase

Properties

InChI Key   BBMHARZCALWXSL-UHFFFAOYSA-M
grade   purum p.a.
assay   ≥99.0% (T)
pH   4.0-4.5 (25 °C, 50 mg/mL in H2O)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S3522 Sodium phosphate monobasic monohydrate, BioReagent, suitable for electrophoresis, 98.0-102.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?