• USA Home
  • 71512 - Sodium dimethyldithiocarbamate hydrate

EMAIL THIS PAGE TO A FRIEND
71512 Sigma-Aldrich

Sodium dimethyldithiocarbamate hydrate

purum, ≥97.0% (NT), 1-2 mol/mol water

Synonym: Dimethyldithiocarbamic acid sodium salt

  • CAS Number 207233-95-2

  • Linear Formula (CH3)2NCSSNa · aq

  • Molecular Weight 143.21 (anhydrous basis)

  •  Beilstein Registry Number 3569024

  •  EC Number 204-876-7

  •  MDL number MFCD00150728

  •  PubChem Substance ID 329762944

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocarbonyl Compounds More...
InChI Key   RJCVAPZBRKHUSV-UHFFFAOYSA-M
grade   purum
assay   ≥97.0% (NT)
impurities   1-2 mol/mol water
mp   120-122 °C (dec.)(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
FD3500000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?