• USA Home
  • 71523 - Sodium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
71523 Sigma-Aldrich

Sodium tetrafluoroborate

purum p.a., ≥97.0% (T)

Synonym: Sodium fluoroborate

Purchase

Properties

Related Categories Essential Chemicals, Research Essentials, Salts, Sodium Salts
InChI Key   KGJZTOFHXCFQIV-UHFFFAOYSA-N
grade   purum p.a.
assay   ≥97.0% (T)
solubility   water: soluble(lit.)
density   2.47 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤1000 mg/kg
  sulfate (SO42-): ≤1000 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability


Laboratory Gloves

Redi-Dri
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?