• USA Home
  • 71596 - Sodium hexafluorosilicate

EMAIL THIS PAGE TO A FRIEND
71596 Sigma-Aldrich

Sodium hexafluorosilicate

for fusion decomposition (of silicates)

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Application Specific Reagents, Decomposition Reagents,
grade   for fusion decomposition (of silicates)
InChI Key   TWGUZEUZLCYTCG-UHFFFAOYSA-N
assay   97%
density   2.68 g/mL at 25 °C(lit.)
cation traces   Ag: ≤100 mg/kg
  Al: ≤100 mg/kg
  Ba: ≤100 mg/kg
  Bi: ≤100 mg/kg
  Ca: ≤100 mg/kg
  Cd: ≤100 mg/kg
  Co: ≤100 mg/kg
  Cr: ≤100 mg/kg
  Cu: ≤100 mg/kg
  Fe: ≤100 mg/kg
  K: ≤100 mg/kg
  Li: ≤100 mg/kg
  Mg: ≤100 mg/kg
  Mn: ≤100 mg/kg
  Mo: ≤100 mg/kg
  Ni: ≤100 mg/kg
  Pb: ≤100 mg/kg
  Sr: ≤100 mg/kg
  Tl: ≤100 mg/kg
  Zn: ≤100 mg/kg

Description

Other Notes

Reagent for the decomposition of silicates by fusion

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2674 6.1 / PGIII
WGK Germany 
2
RTECS 
VV8410000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?