• USA Home
  • 71639 - Sodium phosphate dibasic

EMAIL THIS PAGE TO A FRIEND
71639 Sigma-Aldrich

Sodium phosphate dibasic

BioUltra, anhydrous, ≥99.5% (T)

Synonym: sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Purchase

Properties

Related Categories Alphabetical, BioUltra Buffers, Biochemicals and Reagents, Biological Buffers
grade   anhydrous
InChI Key   BNIILDVGGAEEIG-UHFFFAOYSA-L
product line   BioUltra
assay   ≥99.5% (T)
form   powder or crystals
impurities   insoluble matter, passes filter test
  ≤0.001% total nitrogen (N)
loss   ≤0.1% loss on drying, 110 °C
pH   8.8-9.4 (25 °C, 0.5 M in H2O)
pKa (25 °C)   (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)
solubility   H2O: soluble0.5 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤10 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.2 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤10 mg/kg
  Zn: ≤5 mg/kg
λ   0.5 M in H2O
UV absorption   λ: 260 nm Amax: ≤0.025
  λ: 280 nm Amax: ≤0.020

Description

Other Notes

Primary standard buffer

This product has been replaced by 71636-SIGMA | Sodium phosphate dibasic BioUltra, for molecular biology, ≥99.5% (T)

Price and Availability


Interactive Buffer Table
Safety & Documentation

Safety Information

RIDADR 
UN 3077 9 / PGIII
WGK Germany 
1
RTECS 
WC4500000
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

71636 Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?