• USA Home
  • 71644 - Sodium phosphate dibasic dihydrate

EMAIL THIS PAGE TO A FRIEND
71644 Sigma-Aldrich

Sodium phosphate dibasic dihydrate

BioUltra, ≥99.0% (T)

Synonym: sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate

Purchase

Properties

Related Categories Alphabetical, BioUltra Buffers, Biochemicals and Reagents, Biological Buffers
InChI Key   KDQPSPMLNJTZAL-UHFFFAOYSA-L
product line   BioUltra
assay   ≥99.0% (T)
impurities   insoluble matter, passes filter test
  ≤0.001% total nitrogen (N)
pH   8.9-9.2 (25 °C, 0.5 M in H2O)
solubility   H2O: soluble0.5 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤10 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   0.5 M in H2O
UV absorption   λ: 260 nm Amax: 0.03
  λ: 280 nm Amax: 0.03

Price and Availability


Laboratory Gloves

Interactive Buffer Table
Safety & Documentation

Safety Information

RIDADR 
UN 3077 9 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

71643 Sodium phosphate dibasic dihydrate, BioUltra, for molecular biology, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?