71690 Sigma-Aldrich

Sodium hydroxide

puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets

DOWNLOAD MSDS (PDF)

Synonym: ‘Caustic soda’

  • CAS Number 1310-73-2

  • Linear Formula NaOH

  • Molecular Weight 40.00

  •  EC Number 215-185-5

  •  MDL number MFCD00003548

  •  PubChem Substance ID 24886026

Properties

Related Categories ACS Grade, ACS Grade Acids & Bases, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use,
grade   ACS reagent
vapor density   >1 (vs air)
vapor pressure   <18 mmHg ( 20 °C)
  3 mmHg ( 37 °C)
grade   puriss. p.a.
assay   ≥98.0% (T)
form   pellets
impurities   ≤0.0003% total nitrogen (N)
  ≤0.0005% silicon dioxide (SiO2)
  ≤1.0% sodium carbonate (Na2CO3)
mp   318 °C(lit.)
anion traces   chloride (Cl-): ≤10 mg/kg
  phosphate (PO43-): ≤5 mg/kg
  sulfate (SO42-): ≤5 mg/kg
cation traces   Ag: ≤5 mg/kg
  Al: ≤5 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤5 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  Hg: ≤0.1 mg/kg
  K: ≤200 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Tl: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

Reagent for the decomposition of silicates by fusion1,2

Price and Availability

Customers Also Viewed

Sodium hydroxide

BioXtra, ≥98% (acidimetric), pellets (anhydrous)

Sodium hydroxide

pellets, semiconductor grade, 99.99% trace metals basis

Sodium hydroxide

reagent grade, ≥98%, pellets (anhydrous)

Sodium hydroxide

ACS reagent, ≥97.0%, pellets

Sodium hydroxide

puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?