EMAIL THIS PAGE TO A FRIEND
71700 Sigma-Aldrich

Sodium hydrosulfite

technical, ≥80% (RT)

Synonym: Sodium dithionite, Sodium hypodisulfite

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Inorganic Salts, Research Essentials, Salts,
InChI Key   JVBXVOWTABLYPX-UHFFFAOYSA-L
grade   technical
assay   ≥80% (RT)
solubility   water: soluble(lit.)

Description

Other Notes

Reagent for the reduction of aldehydes and ketones to alcohols; review

Price and Availability


Laboratory Gloves

Redi-Dri
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates toxic gas.
RIDADR 
UN 1384 4.2 / PGII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?