EMAIL THIS PAGE TO A FRIEND
71701 Sigma-Aldrich

Sodium iodate

BioUltra, ≥99.5% (RT)

Purchase

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Sodium, Synthetic Reagents
InChI Key   WTCBONOLBHEDIL-UHFFFAOYSA-M
product line   BioUltra
assay   ≥99.5% (RT)
impurities   insoluble matter, passes filter test
  ≤0.005% total nitrogen (N)
pH   5.5-7.0 (25 °C, 0.1 M in H2O)
solubility   H2O: soluble0.1 M at 20 °C, clear, colorless
density   4.28 g/mL at 25 °C(lit.)
anion traces   bromide, bromate, chloride, chlorate (as Cl-): ≤200 mg/kg
  iodide (I-): ≤20 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤100 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  K: ≤50 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
absorption   absorption0.1M/275 nm, H2O cut-off

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1479 5.1 / PGII
WGK Germany 
3
RTECS 
NN1400000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S4007 Sodium iodate, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?