• USA Home
  • 71748 - Sodium methoxide solution

EMAIL THIS PAGE TO A FRIEND
71748 Sigma-Aldrich

Sodium methoxide solution

technical, ~30% in methanol (T, ~5.4 M)

Synonym: Sodium methylate

  • CAS Number 124-41-4

  • Linear Formula CH3ONa

  • Molecular Weight 54.02

  •  Beilstein Registry Number 3592982

  •  MDL number MFCD00012179

  •  PubChem Substance ID 329763495

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Essential Chemicals, Organic Bases,
InChI Key   WQDUMFSSJAZKTM-UHFFFAOYSA-N
grade   technical
concentration   ~30% in methanol (T, ~5.4 M)
density   0.97 g/mL at 20 °C

Description

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN1289 - class 3 - PG 3 - Sodium methylate solutions
WGK Germany 
1
Flash Point(F) 
60.8 °F
Flash Point(C) 
16 °C
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

156256 Sodium methoxide solution, 25 wt. % in methanol

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?