EMAIL THIS PAGE TO A FRIEND
71799 Sigma-Aldrich

Sodium oxalate

BioUltra, ≥99.5% (RT)

Synonym: Ethanedioic acid sodium salt, Oxalic acid disodium salt

  • CAS Number 62-76-0

  • Linear Formula NaOOCCOONa

  • Molecular Weight 134.00

  •  Beilstein Registry Number 3631622

  •  EC Number 200-550-3

  •  MDL number MFCD00012465

  •  PubChem Substance ID 329763540

Purchase

Properties

Related Categories Alphabetical, BioUltra Buffers, Biochemicals and Reagents, Biological Buffers, Chelators,
InChI Key   ZNCPFRVNHGOPAG-UHFFFAOYSA-L
product line   BioUltra
assay   ≥99.5% (RT)
impurities   insoluble matter, passes filter test
  ≤0.0005% total nitrogen (N)
pH   7-8.5 (25 °C, 0.1 M in H2O)
solubility   H2O: soluble0.1 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤20 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤100 mg/kg
  Cd: ≤10 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  K: ≤50 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤10 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
absorption   absorption0.1M/270 nm, H2O cut-off

Description

Other Notes

Component in a selective media for Yersinia enterocolitica to suppress the growth of gram negative rods

Price and Availability


Laboratory Gloves

Interactive Buffer Table
Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
KI1750000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?