EMAIL THIS PAGE TO A FRIEND
71964 Sigma-Aldrich

Sodium sulfate

purum, anhydrous, ≥99.0% (T)

Purchase

Properties

InChI Key   PMZURENOXWZQFD-UHFFFAOYSA-L
grade   purum
assay   ≥99.0% (T)
mp   884 °C(lit.)
density   2.68 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Sodium sulfate (Na2SO4) may be used in the production of roasted phosphate fertilizers.

General description

Sodium sulfate (Na2SO4) can be obtained directly via precipitation from a saturated solution of Na2SO4 at room temperature (20°C). Its crystallization and growth rates at room temperature (20°C) under various conditions have been investigated.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WE1650000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?