• USA Home
  • 71971 - Sodium sulfate decahydrate

EMAIL THIS PAGE TO A FRIEND
71971 Sigma-Aldrich

Sodium sulfate decahydrate

tested according to Ph.Eur.

Synonym: Glauber’s salt, Natrii sulfas decahydricus

Purchase

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Sodium, Synthetic Reagents
InChI Key   RSIJVJUOQBWMIM-UHFFFAOYSA-L
pharmacopeia   testing conforms to Pharmacopeia
density   1.46 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?