72049 Sigma-Aldrich

Sodium thiosulfate

purum p.a., anhydrous, ≥98.0% (RT)

DOWNLOAD MSDS (PDF)
  • CAS Number 7772-98-7

  • Linear Formula Na2S2O3

  • Molecular Weight 158.11

  •  EC Number 231-867-5

  •  MDL number MFCD00003499

  •  PubChem Substance ID 24886350

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Sodium, Synthetic Reagents
grade   purum p.a.
assay   ≥98.0% (RT)
pH   6.0-8.5 (25 °C, 50 mg/mL in H2O)
anion traces   sulfide (S2-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤500 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability

Customers Also Viewed

Potassium iodide

puriss. p.a., ACS reagent, ≥99.0% (AT)

Sodium thiosulfate

≥99.99% trace metals basis

Sodium thiosulfate

ReagentPlus®, 99%

Sodium thiosulfate solution

2 g/dL in deionized water

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?