• USA Home
  • 72062 - Sodium triacetoxyborohydride

EMAIL THIS PAGE TO A FRIEND
72062 Sigma-Aldrich

Sodium triacetoxyborohydride

technical, ≥90% (gas-volumetric)

Synonym: STAB

  • CAS Number 56553-60-7

  • Linear Formula (CH3COO)3BHNa

  • Molecular Weight 211.94

  •  Beilstein Registry Number 4047608

  •  MDL number MFCD00012211

  •  PubChem Substance ID 24886353

Purchase

Properties

Related Categories Borohydrides, Chemical Synthesis, Reduction, Synthetic Reagents
InChI Key   HHYFEYBWNZJVFQ-UHFFFAOYSA-N
grade   technical
assay   ≥90% (gas-volumetric)
mp   116-120 °C (dec.)(lit.)

Description

Other Notes

Mild, stereoselective reducing agent

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 1409 4.3 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?