• USA Home
  • 72065 - Sodium bismuthate(V) hydrate

EMAIL THIS PAGE TO A FRIEND
72065 Sigma-Aldrich

Sodium bismuthate(V) hydrate

for the determ. of manganese in iron and steel, oxidizing agent, ≥85% (RT)

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Application Specific Reagents, Decomposition Reagents,
grade   oxidizing agent
InChI Key   UQGDUPFKOOSTOS-UHFFFAOYSA-N
assay   ≥85% (RT)
quality   for the determ. of manganese in iron and steel
impurities   ≤0.008% soluble peroxides (as O)
anion traces   chloride (Cl-): ≤20 mg/kg
  nitrate (NO3-): ≤40 mg/kg
cation traces   Mn: ≤5 mg/kg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
VZ1750000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?